Assay Design



Gene Transcripts

Gene Symbol
AQP11
Entrez Gene ID
102146208
Gene Name
aquaporin 11
Gene Aliases
-
Chromosome Location
Chr.14: 72924771 - 72945228 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579170.2 XP_005579227.1 1 - 2 938 80

View Full Product Details