Assay Design



Gene Transcripts

Gene Symbol
SLC18A3
Entrez Gene ID
102147099
Gene Name
solute carrier family 18 member A3
Gene Aliases
-
Chromosome Location
Chr.9: 45459253 - 45461665 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005565137.1 XP_005565194.1 1 - 1 2160 117

View Full Product Details