Assay Design



Gene Transcripts

Gene Symbol
FOXE1
Entrez Gene ID
102125419
Gene Name
forkhead box E1
Gene Aliases
-
Chromosome Location
Chr.15: 39922027 - 39927034 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015436686.1 XP_015292172.1 2 - 2 2144 79

View Full Product Details