Assay Design



Gene Transcripts

Gene Symbol
COL8A2
Entrez Gene ID
102143193
Gene Name
collagen type VIII alpha 2 chain
Gene Aliases
-
Chromosome Location
Chr.1: 191763826 - 191768909 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015438407.1 XP_015293893.1 1 - 2 301 105

View Full Product Details