Assay Design



Gene Transcripts

Gene Symbol
LBX1
Entrez Gene ID
102136423
Gene Name
ladybird homeobox 1
Gene Aliases
-
Chromosome Location
Chr.9: 100185726 - 100188460 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015456917.1 XP_015312403.1 1 - 2 466 85

View Full Product Details