Assay Design



Gene Transcripts

Gene Symbol
PCDHB1
Entrez Gene ID
102135968
Gene Name
protocadherin beta 1
Gene Aliases
-
Chromosome Location
Chr.6: 140179960 - 140185296 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558010.2 XP_005558067.1 1 - 1 2244 164

View Full Product Details