Assay Design



Gene Transcripts

Gene Symbol
SNRPD2
Entrez Gene ID
102124807
Gene Name
small nuclear ribonucleoprotein D2 polypeptide
Gene Aliases
-
Chromosome Location
Chr.19: 46506401 - 46510944 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589608.2 XP_005589665.1 1 - 2 302 123

View Full Product Details