Assay Design



Gene Transcripts

Gene Symbol
LLPH
Entrez Gene ID
102119216
Gene Name
LLP homolog, long-term synaptic facilitation factor
Gene Aliases
-
Chromosome Location
Chr.11: 65197090 - 65204929 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005571460.2 XP_005571517.1 3 - 3 356 135

View Full Product Details