Assay Design



Gene Transcripts

Gene Symbol
LOC102145420
Entrez Gene ID
102145420
Gene Name
gamma-crystallin C
Gene Aliases
CRYGC
Chromosome Location
Chr.12: 97746039 - 97747781 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005574095.2 XP_005574152.1 2 - 3 334 73
XM_015432675.1 XP_015288161.1 1 - 2 326 73

View Full Product Details