Assay Design



Gene Transcripts

Gene Symbol
PEG10
Entrez Gene ID
102125830
Gene Name
paternally expressed 10
Gene Aliases
-
Chromosome Location
Chr.3: 117319438 - 117332767 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291293.1 NP_001278222.1 2 - 2 2919 93
NM_001291294.1 NP_001278223.1 2 - 2 2919 93

View Full Product Details