Assay Design



Gene Transcripts

Gene Symbol
TCEAL7
Entrez Gene ID
102119599
Gene Name
transcription elongation factor A like 7
Gene Aliases
-
Chromosome Location
Chr.X: 100155817 - 100157969 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005594223.2 XP_005594280.1 1 - 2 130 93

View Full Product Details