Assay Design



Gene Transcripts

Gene Symbol
TCEAL7
Entrez Gene ID
102119599
Gene Name
transcription elongation factor A like 7
Gene Aliases
-
Chromosome Location
Chr.X: 100155817 - 100157969 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005594223.2 XP_005594280.1 2 - 3 189 76
XM_005594224.1 XP_005594281.1 2 - 3 229 76
XM_015443808.1 XP_015299294.1 2 - 3 85 76

View Full Product Details