Assay Design



Gene Transcripts

Gene Symbol
LOC102145084
Entrez Gene ID
102145084
Gene Name
serine/arginine-rich splicing factor 8
Gene Aliases
SRSF8
Chromosome Location
Chr.14: 90949290 - 90953608 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579398.2 XP_005579455.1 1 - 1 991 56

View Full Product Details