Assay Design



Gene Transcripts

Gene Symbol
IGSF6
Entrez Gene ID
102139981
Gene Name
immunoglobulin superfamily member 6
Gene Aliases
-
Chromosome Location
Chr.20: 21342032 - 21356121 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283642.1 NP_001270571.1 1 - 2 97 75
XM_005591446.2 XP_005591503.1 1 - 2 75
GenBank mRNA AB168410.1 - 1 - 2 97 75

View Full Product Details