Assay Design



Gene Transcripts

Gene Symbol
MGAT4C
Entrez Gene ID
101866242
Gene Name
MGAT4 family member C
Gene Aliases
GnT-IVc
Chromosome Location
Chr.11: 85289196 - 85783326 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283085.1 NP_001270014.1 5 - 6 621 118
XM_015430648.1 XP_015286134.1 6 - 7 1290 118
XM_015430649.1 XP_015286135.1 6 - 7 1332 118
XM_015430650.1 XP_015286136.1 6 - 7 1285 118
XM_015430651.1 XP_015286137.1 6 - 7 767 118
XM_015430652.1 XP_015286138.1 4 - 5 1065 118
XM_015430653.1 XP_015286139.1 4 - 5 833 118
GenBank mRNA AB179007.1 - 5 - 6 621 118

View Full Product Details