Assay Design



Gene Transcripts

Gene Symbol
YIPF5
Entrez Gene ID
101866225
Gene Name
Yip1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.6: 143350136 - 143363867 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285289.1 NP_001272218.1 4 - 5 534 87
XM_005558118.2 XP_005558175.1 4 - 5 884 87
GenBank mRNA AB171857.1 - 4 - 5 534 87

View Full Product Details