Assay Design



Gene Transcripts

Gene Symbol
MED10
Entrez Gene ID
101925797
Gene Name
mediator complex subunit 10
Gene Aliases
-
Chromosome Location
Chr.6: 6377233 - 6383836 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284680.1 NP_001271609.1 1 - 2 188 76
GenBank mRNA AB169066.1 - 1 - 2 188 76

View Full Product Details