Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001283096.1 | NP_001270025.1 | 11 - 12 | 2096 | 108 |
XM_005550638.2 | XP_005550695.1 | 12 - 13 | 2625 | 108 | |
XM_005550639.2 | XP_005550696.1 | 10 - 11 | 2426 | 108 | |
XM_005550640.2 | XP_005550697.1 | 10 - 11 | 2423 | 108 | |
XM_015447303.1 | XP_015302789.1 | 11 - 12 | 2031 | 108 | |
GenBank mRNA | AB169093.1 | - | 11 - 12 | 2096 | 108 |
AB169129.1 | - | 9 - 10 | 1493 | 108 |