Assay Design



Gene Transcripts

Gene Symbol
SPATA16
Entrez Gene ID
101926490
Gene Name
spermatogenesis associated 16
Gene Aliases
-
Chromosome Location
Chr.18: 69729054 - 69745469 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283746.1 NP_001270675.1 - 1334 63
GenBank mRNA AB179163.1 - - 1334 63

View Full Product Details