Assay Design



Gene Transcripts

Gene Symbol
AVPI1
Entrez Gene ID
102127834
Gene Name
arginine vasopressin induced 1
Gene Aliases
-
Chromosome Location
Chr.9: 96501249 - 96511519 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566133.2 XP_005566190.1 2 - 3 977 85

View Full Product Details