Assay Design



Gene Transcripts

Gene Symbol
HOXB2
Entrez Gene ID
102129644
Gene Name
homeobox B2
Gene Aliases
-
Chromosome Location
Chr.16: 33896272 - 33899079 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015438215.1 XP_015293701.1 1 - 2 923 92

View Full Product Details