Assay Design



Gene Transcripts

Gene Symbol
CEACAM5
Entrez Gene ID
102128791
Gene Name
CEA cell adhesion molecule 5
Gene Aliases
-
Chromosome Location
Chr.19: 42218251 - 42238929 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589434.2 XP_005589491.1 4 - 5 1103 66

View Full Product Details