Assay Design



Gene Transcripts

Gene Symbol
HSPB9
Entrez Gene ID
102115562
Gene Name
heat shock protein family B (small) member 9
Gene Aliases
-
Chromosome Location
Chr.16: 53831799 - 53833018 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005596051.2 XP_005596108.1 1 - 1 1154 69

View Full Product Details