Assay Design



Gene Transcripts

Gene Symbol
OXSM
Entrez Gene ID
102127446
Gene Name
3-oxoacyl-ACP synthase, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.2: 21621757 - 21633256 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005545626.2 XP_005545683.1 3 - 4 1424 70
XM_015445021.1 XP_015300507.1 4 - 5 1973 70
XM_015445022.1 XP_015300508.1 4 - 5 1805 70

View Full Product Details