Assay Design



Gene Transcripts

Gene Symbol
PYY
Entrez Gene ID
102136928
Gene Name
peptide YY
Gene Aliases
-
Chromosome Location
Chr.16: 55707375 - 55711623 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005584388.2 XP_005584445.2 2 - 3 289 84
XM_015438560.1 XP_015294046.1 3 - 4 2412 84
XM_015438561.1 XP_015294047.1 3 - 4 2414 84

View Full Product Details