Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015438823.1 | XP_015294309.1 | 4 - 5 | 2430 | 101 |
XM_015438824.1 | XP_015294310.1 | 5 - 6 | 1707 | 101 | |
XM_015438825.1 | XP_015294311.1 | 5 - 6 | 1670 | 101 | |
XM_015438826.1 | XP_015294312.1 | 5 - 6 | 1476 | 101 | |
XM_015438827.1 | XP_015294313.1 | 4 - 5 | 1230 | 101 | |
XM_015438829.1 | XP_015294315.1 | 4 - 5 | 1233 | 101 |