Assay Design



Gene Transcripts

Gene Symbol
TREML4
Entrez Gene ID
102130989
Gene Name
triggering receptor expressed on myeloid cells like 4
Gene Aliases
-
Chromosome Location
Chr.4: 129440608 - 129451386 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005595974.2 XP_005596031.2 3 - 4 527 110
XM_015449422.1 XP_015304908.1 3 - 4 527 110

View Full Product Details