Assay Design



Gene Transcripts

Gene Symbol
PGPEP1L
Entrez Gene ID
102131928
Gene Name
pyroglutamyl-peptidase I like
Gene Aliases
-
Chromosome Location
Chr.7: 79773146 - 79777374 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015453440.1 XP_015308926.1 2 - 3 786 88

View Full Product Details