Assay Design



Gene Transcripts

Gene Symbol
STUM
Entrez Gene ID
102116807
Gene Name
stum, mechanosensory transduction mediator homolog
Gene Aliases
-
Chromosome Location
Chr.1: 85409407 - 85474121 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541013.2 XP_005541070.1 2 - 3 464 95

View Full Product Details