Assay Design



Gene Transcripts

Gene Symbol
TNFAIP8L3
Entrez Gene ID
102117064
Gene Name
TNF alpha induced protein 8 like 3
Gene Aliases
-
Chromosome Location
Chr.7: 28042353 - 28090875 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005559553.2 XP_005559610.1 2 - 3 508 100
XM_005559554.2 XP_005559611.1 1 - 2 187 100

View Full Product Details