Assay Design



Gene Transcripts

Gene Symbol
ICAM4
Entrez Gene ID
102139593
Gene Name
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
Gene Aliases
-
Chromosome Location
Chr.19: 10563626 - 10565347 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005587936.2 XP_005587993.1 1 - 2 544 60

View Full Product Details