Assay Design



Gene Transcripts

Gene Symbol
NACC1
Entrez Gene ID
102126978
Gene Name
nucleus accumbens associated 1
Gene Aliases
-
Chromosome Location
Chr.19: 13357247 - 13376142 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005588190.2 XP_005588247.1 3 - 4 411 117

View Full Product Details