Assay Design



Gene Transcripts

Gene Symbol
ADGRE3
Entrez Gene ID
102118304
Gene Name
adhesion G protein-coupled receptor E3
Gene Aliases
EMR3
Chromosome Location
Chr.19: 15011547 - 15079187 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015440203.1 XP_015295689.1 13 - 14 2229 109

View Full Product Details