Assay Design



Gene Transcripts

Gene Symbol
AADACL3
Entrez Gene ID
102141239
Gene Name
arylacetamide deacetylase like 3
Gene Aliases
-
Chromosome Location
Chr.1: 215345461 - 215358817 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005544757.2 XP_005544814.1 2 - 3 941 86
XM_015442556.1 XP_015298042.1 3 - 4 724 86

View Full Product Details