Assay Design



Gene Transcripts

Gene Symbol
MANSC4
Entrez Gene ID
102126872
Gene Name
MANSC domain containing 4
Gene Aliases
-
Chromosome Location
Chr.11: 28450634 - 28459937 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570452.2 XP_005570509.2 1 - 2 763 105

View Full Product Details