Assay Design



Gene Transcripts

Gene Symbol
VGLL1
Entrez Gene ID
102137749
Gene Name
vestigial like family member 1
Gene Aliases
-
Chromosome Location
Chr.X: 133299994 - 133336438 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015444336.1 XP_015299822.1 2 - 3 320 66
XM_015444337.1 XP_015299823.1 1 - 2 231 66

View Full Product Details