Assay Design



Gene Transcripts

Gene Symbol
TNFRSF13C
Entrez Gene ID
102116472
Gene Name
TNF receptor superfamily member 13C
Gene Aliases
-
Chromosome Location
Chr.10: 9146357 - 9148440 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005567127.2 XP_005567184.1 2 - 3 442 59

View Full Product Details