Assay Design



Gene Transcripts

Gene Symbol
LOC102129427
Entrez Gene ID
102129427
Gene Name
leukocyte immunoglobulin-like receptor subfamily B member 5
Gene Aliases
LILRB5
Chromosome Location
Chr.19: 55132391 - 55139221 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590293.2 XP_005590350.1 9 - 10 1584 53
XM_005590297.2 XP_005590354.1 9 - 10 1584 53

View Full Product Details