Assay Design



Gene Transcripts

Gene Symbol
TYSND1
Entrez Gene ID
102135762
Gene Name
trypsin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 65534101 - 65542131 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005565649.2 XP_005565706.1 4 - 5 1550 62

View Full Product Details