Assay Design



Gene Transcripts

Gene Symbol
MS4A15
Entrez Gene ID
102116162
Gene Name
membrane spanning 4-domains A15
Gene Aliases
-
Chromosome Location
Chr.14: 13495347 - 13515393 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577703.2 XP_005577760.1 4 - 5 1360 72

View Full Product Details