Assay Design



Gene Transcripts

Gene Symbol
HS3ST3A1
Entrez Gene ID
102131170
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 3A1
Gene Aliases
-
Chromosome Location
Chr.16: 13705531 - 13807370 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005582937.1 XP_005582994.1 1 - 2 1394 65

View Full Product Details