Assay Design



Gene Transcripts

Gene Symbol
CTSG
Entrez Gene ID
102128983
Gene Name
cathepsin G
Gene Aliases
-
Chromosome Location
Chr.7: 88345329 - 88348096 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005560993.1 XP_005561050.1 2 - 3 246 71

View Full Product Details