Assay Design



Gene Transcripts

Gene Symbol
LOC102141319
Entrez Gene ID
102141319
Gene Name
lithostathine-1-beta
Gene Aliases
-
Chromosome Location
Chr.13: 30589195 - 30591581 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005575468.2 XP_005575525.1 4 - 5 431 78

View Full Product Details