Assay Design



Gene Transcripts

Gene Symbol
FOXR1
Entrez Gene ID
102122342
Gene Name
forkhead box R1
Gene Aliases
-
Chromosome Location
Chr.14: 114997473 - 115009374 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579866.2 XP_005579923.1 2 - 3 716 86
XM_015435820.1 XP_015291306.1 2 - 3 227 86

View Full Product Details