Assay Design



Gene Transcripts

Gene Symbol
SPRYD4
Entrez Gene ID
102134168
Gene Name
SPRY domain containing 4
Gene Aliases
-
Chromosome Location
Chr.11: 55428849 - 55431353 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005571236.2 XP_005571293.1 1 - 2 187 99

View Full Product Details