Assay Design



Gene Transcripts

Gene Symbol
TGOLN2
Entrez Gene ID
102123547
Gene Name
trans-golgi network protein 2
Gene Aliases
TGN47
Chromosome Location
Chr.13: 24126120 - 24138825 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015433489.1 XP_015288975.1 2 - 3 1220 83
XM_015433490.1 XP_015288976.1 2 - 3 1220 83
GenBank mRNA X94334.1 - - 1267 83

View Full Product Details