Assay Design



Gene Transcripts

Gene Symbol
GBP7
Entrez Gene ID
102145455
Gene Name
guanylate binding protein 7
Gene Aliases
-
Chromosome Location
Chr.1: 138057401 - 138103156 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005542797.2 XP_005542854.2 5 - 6 795 75

View Full Product Details