Assay Design



Gene Transcripts

Gene Symbol
CRYGN
Entrez Gene ID
102126237
Gene Name
crystallin gamma N
Gene Aliases
-
Chromosome Location
Chr.3: 184136494 - 184149321 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005551215.2 XP_005551272.1 3 - 4 666 70
XM_015448305.1 XP_015303791.1 3 - 4 864 70

View Full Product Details