Assay Design



Gene Transcripts

Gene Symbol
TPTE2
Entrez Gene ID
102136934
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
Gene Aliases
-
Chromosome Location
Chr.17: 1874522 - 1971908 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015439049.1 XP_015294535.1 21 - 22 2085 122
GenBank mRNA AB169152.1 - 11 - 12 1040 122

View Full Product Details