Assay Design



Gene Transcripts

Gene Symbol
TWIST1
Entrez Gene ID
102120863
Gene Name
twist family bHLH transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.3: 101618215 - 101620805 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015447650.1 XP_015303136.1 2 - 3 771 91

View Full Product Details